1-Bromo-3-chloro-2-fluoro-4-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

112083-500MG 112083-1G 112083-5G 112083-25G
77323-490EA 215.27 USD
77323-490 77347-554 77359-600 77383-486
1-Bromo-3-chloro-2-fluoro-4-methoxybenzene ≥95%
1-Bromo-3-chloro-2-fluoro-4-methoxybenzene

Precaución: for Research Use Only

Formula: C₇H₅BrClFO
MW: 239.469 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18782091
CAS Number: 909122-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR