2-Bromopivalanilide ≥97%

Supplier: AOBChem USA

Warning

11187-500MG 11187-1G 11187-5G 11187-25G
77323-488EA 121.54 USD
77323-488 77343-902 77359-598 77371-576
2-Bromopivalanilide ≥97%
2-Bromopivalanilide

Precaución: for Research Use Only

Formula: C₁₁H₁₄BrNO
Storage Temperature: Ambient
MDL Number: MFCD00507417
CAS Number: 65854-92-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR