1-(2-(3-fluoro-2-methylphenoxy)ethyl)piperidine ≥97%

Supplier: AOBChem USA

Warning

111775-500MG 111775-1G 111775-5G 111775-25G
77323-486EA 264.71 USD
77323-486 77347-552 77359-596 77383-484
1-(2-(3-fluoro-2-methylphenoxy)ethyl)piperidine ≥97%
1-(2-(3-fluoro-2-methylphenoxy)ethyl)piperidine

Precaución: for Research Use Only

Formula: C₁₄H₂₀FNO
MW: 237.313 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34781224
CAS Number: 2918881-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR