4-Bromo-3-chloro-2-methylanisole ≥97%

Supplier: AOBChem USA

Warning

11159-500MG 11159-1G 11159-5G 11159-25G
77323-484EA 244.11 USD
77323-484 77347-550 77359-594 77383-482
4-Bromo-3-chloro-2-methylanisole ≥97%
4-Bromo-3-chloro-2-methylanisole

Precaución: for Research Use Only

Formula: C₈H₈BrClO
MW: 235.506 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22414516
CAS Number: 1155264-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR