1-Bromo-5-chloro-2-ethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

11141-1G 11141-5G 11141-25G
77340-582EA 60.77 USD
77340-582 77359-592 77371-572
1-Bromo-5-chloro-2-ethoxybenzene ≥97%
1-Bromo-5-chloro-2-ethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₈BrClO
Storage Temperature: Ambient
MDL Number: MFCD16318792
CAS Number: 1225577-71-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR