6,7-Dibromo-1H-indazole ≥95%

Supplier: AOBChem USA

Warning

111352-500MG 111352-1G 111352-5G
77340-042EA 722.03 USD
77340-042 77347-548 77371-570
6,7-Dibromo-1H-indazole ≥95%
6,7-Dibromo-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄Br₂N₂
MW: 275.928 g/mol
Storage Temperature: Ambient
CAS Number: 2384019-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR