1-Fluoro-2,3-diiodo-4-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

111218-500MG 111218-1G 111218-5G
77339-856EA 662.29 USD
77339-856 77347-544 77371-566
1-Fluoro-2,3-diiodo-4-methoxybenzene ≥97%
1-Fluoro-2,3-diiodo-4-methoxybenzene

Precaución: for Research Use Only

Formula: C₇H₅FI₂O
MW: 377.921 g/mol
Storage Temperature: Ambient
CAS Number: 1806298-86-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR