Trimethyl(2-(trifluoromethyl)phenyl)silane ≥97%

Supplier: AOBChem USA

Warning

110900-500MG 110900-1G 110900-5G 110900-25G
77323-478EA 202.91 USD
77323-478 77347-540 77359-588 77383-480
Trimethyl(2-(trifluoromethyl)phenyl)silane ≥97%
Trimethyl(2-(trifluoromethyl)phenyl)silane

Precaución: for Research Use Only

Formula: C₁₀H₁₃F₃Si
MW: 218.291 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30178939
CAS Number: 312-92-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR