4-Benzyloxy-3-chloro-5-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

11078-500MG 11078-1G 11078-5G 11078-25G
77323-476EA 121.54 USD
77323-476 77343-900 77359-586 77371-558
4-Benzyloxy-3-chloro-5-methylphenylboronic acid ≥97%
4-Benzyloxy-3-chloro-5-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₄BClO₃
MW: 276.523 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04039001
CAS Number: 1050509-78-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR