3-((2-Fluorobenzyl)oxy)-4-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

110522-1G 110522-5G 110522-25G
77342-066EA 111.24 USD
77342-066 77359-582 77371-554
3-((2-Fluorobenzyl)oxy)-4-methylbenzaldehyde ≥97%
3-((2-Fluorobenzyl)oxy)-4-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₃FO₂
Storage Temperature: Ambient
MDL Number: MFCD34780687
CAS Number: 2918932-67-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR