1-Ethoxy-4-fluoro-2-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

110264-500MG 110264-1G 110264-5G 110264-25G
77323-472EA 115.36 USD
77323-472 77343-408 77359-578 77371-548
1-Ethoxy-4-fluoro-2-isopropoxybenzene ≥97%
1-Ethoxy-4-fluoro-2-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₅FO₂
Storage Temperature: Ambient
MDL Number: MFCD34780500
CAS Number: 2901102-98-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR