1,2-diethoxy-4-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

110263-500MG 110263-1G 110263-5G 110263-25G
77323-470EA 127.72 USD
77323-470 77344-224 77359-576 77371-546
1,2-diethoxy-4-fluorobenzene ≥97%
1,2-diethoxy-4-fluorobenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃FO₂
MW: 184.207 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34780499
CAS Number: 219998-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR