2,4-Dibromo-6-ethoxyphenol ≥97%

Supplier: AOBChem USA

Warning

109678-500MG 109678-1G 109678-5G 109678-25G
77323-462EA 174.07 USD
77323-462 77346-474 77359-566 77383-474
2,4-Dibromo-6-ethoxyphenol ≥97%
2,4-Dibromo-6-ethoxyphenol

Precaución: for Research Use Only

Formula: C₈H₈Br₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34780280
CAS Number: 53948-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR