2-Bromo-6-fluoro-4-methylbenzonitrile ≥95%

Supplier: AOBChem USA

Warning

109593-1G 109593-5G 109593-25G
77342-086EA 112.27 USD
77342-086 77359-564 77371-532
2-Bromo-6-fluoro-4-methylbenzonitrile ≥95%
2-Bromo-6-fluoro-4-methylbenzonitrile

Precaución: for Research Use Only

Formula: C₈H₅BrFN
Storage Temperature: Ambient
MDL Number: MFCD28503349
CAS Number: 1023971-89-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR