1-Chloro-4-isopropyl-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

109365-500MG 109365-1G 109365-5G
77337-684EA 435.69 USD
77337-684 77347-526 77371-530
1-Chloro-4-isopropyl-2-methylbenzene ≥97%
1-Chloro-4-isopropyl-2-methylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃Cl
MW: 168.663 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29093120
CAS Number: 15146-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR