1-(2-Chloro-4-methoxypyridin-3-yl)ethanol ≥97%

Supplier: AOBChem USA

Warning

109359-500MG 109359-1G 109359-25G 109359-5G
77323-458EA 305.91 USD
77323-458 77347-524 77383-470 77371-528
1-(2-Chloro-4-methoxypyridin-3-yl)ethanol ≥97%
1-(2-Chloro-4-methoxypyridin-3-yl)ethanol

Precaución: for Research Use Only

Formula: C₈H₁₀ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD34780131
CAS Number: 1784516-35-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR