2,5-diiodo-1,3-diisopropylbenzene ≥97%

Supplier: AOBChem USA

Warning

108989-500MG 108989-1G 108989-5G
77337-610EA 426.42 USD
77337-610 77347-520 77371-526
2,5-diiodo-1,3-diisopropylbenzene ≥97%
2,5-diiodo-1,3-diisopropylbenzene

Precaución: for Research Use Only

Formula: C₁₂H₁₆I₂
MW: 414.064 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34780049
CAS Number: 2901066-54-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR