4-Iodo-2,6-diisopropylaniline ≥95%

Supplier: AOBChem USA

Warning

108987-500MG 108987-1G 108987-5G
77337-618EA 429.51 USD
77337-618 77347-518 77371-524
4-Iodo-2,6-diisopropylaniline ≥95%
4-Iodo-2,6-diisopropylaniline

Precaución: for Research Use Only

Formula: C₁₂H₁₈IN
Storage Temperature: Ambient
MDL Number: MFCD27965382
CAS Number: 140411-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR