2-Chloro-3-cyanoisonicotinic acid ≥95%

Supplier: AOBChem USA

Warning

108910-500MG 108910-1G 108910-5G 108910-25G
77323-454EA 259.56 USD
77323-454 77347-516 77359-556 77383-466
2-Chloro-3-cyanoisonicotinic acid ≥95%
2-Chloro-3-cyanoisonicotinic acid

Precaución: for Research Use Only

Formula: C₇H₃ClN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD28749703
CAS Number: 1807049-85-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR