8-Chloro-6-methylquinolin-2(1H)-one ≥97%

Supplier: AOBChem USA

Warning

108771-500MG 108771-1G 108771-5G
77335-930EA 368.74 USD
77335-930 77347-514 77371-518
8-Chloro-6-methylquinolin-2(1H)-one ≥97%
8-Chloro-6-methylquinolin-2(1H)-one

Precaución: for Research Use Only

Formula: C₁₀H₈ClNO
Storage Temperature: Ambient
CAS Number: 701291-30-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR