5-Chloro-4-fluoro-2-methoxyaniline ≥97%
Supplier: AOBChem USA
|
Warning
|
108754-500MG
108754-1G
108754-5G
108754-25G
77323-450EA
93.73
USD
77323-450
77342-514
77359-552
77371-514
5-Chloro-4-fluoro-2-methoxyaniline ≥97%
5-Chloro-4-fluoro-2-methoxyaniline
Precaución: for Research Use Only
Formula:
C₇H₇ClFNO Storage Temperature: Ambient |
MDL Number:
MFCD11847614 CAS Number: 1394839-94-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...