1-Chloro-2-fluoro-5-iodo-4-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

108673-500MG 108673-1G 108673-5G 108673-25G
77323-446EA 269.86 USD
77323-446 77347-506 77359-548 77383-462
1-Chloro-2-fluoro-5-iodo-4-methoxybenzene ≥97%
1-Chloro-2-fluoro-5-iodo-4-methoxybenzene

Precaución: for Research Use Only

Formula: C₇H₅ClFIO
MW: 286.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31725574
CAS Number: 2168547-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR