Methyl 5-bromo-3-fluoro-2-hydroxybenzoate ≥95%

Supplier: AOBChem USA

Warning

108616-500MG 108616-1G 108616-5G 108616-25G
77323-442EA 110.21 USD
77323-442 77343-284 77359-544 77371-510
Methyl 5-bromo-3-fluoro-2-hydroxybenzoate ≥95%
Methyl 5-bromo-3-fluoro-2-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₈H₆BrFO₃
Storage Temperature: Ambient
MDL Number: MFCD06204235
CAS Number: 773134-37-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR