4-Bromo-2,3-difluoro-N-methoxy-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

108575-1G 108575-5G 108575-25G
77341-118EA 71.07 USD
77341-118 77359-542 77371-508
4-Bromo-2,3-difluoro-N-methoxy-N-methylbenzamide ≥97%
4-Bromo-2,3-difluoro-N-methoxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₉H₈BrF₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD34779917
CAS Number: 2901105-85-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR