4-Bromo-1-(cyclopropylmethyl)-1H-pyrazole ≥95%

Supplier: AOBChem USA

Warning

108459-1G 108459-5G 108459-25G
77341-074EA 69.01 USD
77341-074 77359-540 77371-504
4-Bromo-1-(cyclopropylmethyl)-1H-pyrazole ≥95%
4-Bromo-1-(cyclopropylmethyl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₇H₉BrN₂
Storage Temperature: Ambient
MDL Number: MFCD12403651
CAS Number: 1216152-26-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR