2-Bromo-3-(cyclopropylmethoxy)pyridine ≥97%

Supplier: AOBChem USA

Warning

108205-500MG 108205-1G 108205-5G 108205-25G
77323-438EA 261.62 USD
77323-438 77347-500 77359-538 77383-458
2-Bromo-3-(cyclopropylmethoxy)pyridine ≥97%
2-Bromo-3-(cyclopropylmethoxy)pyridine

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO
MW: 228.086 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21162928
CAS Number: 910606-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR