2-Bromo-6-iodobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

108111-500MG 108111-1G 108111-5G 108111-25G
77323-434EA 213.21 USD
77323-434 77347-496 77359-534 77383-454
2-Bromo-6-iodobenzaldehyde ≥97%
2-Bromo-6-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄BrIO
MW: 310.915 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11520408
CAS Number: 1261497-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR