6-Bromo-2,3,4-trifluorobenzonitrile ≥97%

Supplier: AOBChem USA

Warning

108027-500MG 108027-1G 108027-25G 108027-5G
77323-432EA 290.46 USD
77323-432 77347-494 77383-452 77371-500
6-Bromo-2,3,4-trifluorobenzonitrile ≥97%
6-Bromo-2,3,4-trifluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇HBrF₃N
MW: 235.989 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32264074
CAS Number: 2407339-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR