2,3-Dichloro-4-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

107993-1G 107993-5G 107993-25G
77341-708EA 100.94 USD
77341-708 77359-530 77371-498
2,3-Dichloro-4-(methylthio)benzoic acid ≥97%
2,3-Dichloro-4-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₆Cl₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD34779669
CAS Number: 105917-74-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR