6-Bromo-7-(methylthio)-2,3-dihydrobenzo[b][1,4]dioxine ≥97%

Supplier: AOBChem USA

Warning

107988-500MG 107988-1G 107988-5G 107988-25G
77323-428EA 142.14 USD
77323-428 77344-994 77359-524 77383-448
6-Bromo-7-(methylthio)-2,3-dihydrobenzo[b][1,4]dioxine ≥97%
6-Bromo-7-(methylthio)-2,3-dihydrobenzo[b][1,4]dioxine

Precaución: for Research Use Only

Formula: C₉H₉BrO₂S
MW: 261.136 g/mol
Storage Temperature: Ambient
CAS Number: 2921886-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR