7-Bromo-2,3-dihydrobenzo[b][1,4]dioxine-6-carboxylic acid ≥95%

Supplier: AOBChem USA

Warning

107986-500MG 107986-1G 107986-5G 107986-25G
77323-426EA 114.33 USD
77323-426 77343-390 77359-522 77371-492
7-Bromo-2,3-dihydrobenzo[b][1,4]dioxine-6-carboxylic acid ≥95%
7-Bromo-2,3-dihydrobenzo[b][1,4]dioxine-6-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₇BrO₄
Storage Temperature: Ambient
MDL Number: MFCD00510294
CAS Number: 59820-91-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR