3,4-Dibromo-5-fluorophenol ≥95%

Supplier: AOBChem USA

Warning

107959-500MG 107959-1G
77340-474EA 1204.07 USD
77340-474 77347-490
3,4-Dibromo-5-fluorophenol ≥95%
3,4-Dibromo-5-fluorophenol

Precaución: for Research Use Only

Formula: C₆H₃Br₂FO
Storage Temperature: Ambient
CAS Number: 1803716-58-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR