1-(Cyclopent-1-en-1-yl)-4-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

107951-500MG 107951-1G
77340-466EA 1186.56 USD
77340-466 77347-488
1-(Cyclopent-1-en-1-yl)-4-fluorobenzene ≥97%
1-(Cyclopent-1-en-1-yl)-4-fluorobenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₁F
MW: 162.203 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11870705
CAS Number: 827-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR