3-Bromo-5-(tert-butyl)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

107531-500MG 107531-1G 107531-5G
77337-242EA 408.91 USD
77337-242 77347-482 77371-486
3-Bromo-5-(tert-butyl)benzonitrile ≥97%
3-Bromo-5-(tert-butyl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₁H₁₂BrN
Storage Temperature: Ambient
MDL Number: MFCD28399387
CAS Number: 1369836-56-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR