4-Bromo-1-tert-butoxy-2-chlorobenzene ≥97%

Supplier: AOBChem USA

Warning

10751-500MG 10751-1G 10751-5G 10751-25G
77323-420EA 101.97 USD
77323-420 77342-976 77359-516 77371-484
4-Bromo-1-tert-butoxy-2-chlorobenzene ≥97%
4-Bromo-1-tert-butoxy-2-chlorobenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrClO
Storage Temperature: Ambient
MDL Number: MFCD18917763
CAS Number: 1309932-97-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR