5-Bromo-4-methoxy-2-methylphenol ≥95%

Supplier: AOBChem USA

Warning

107432-500MG 107432-1G 107432-5G
77339-956EA 692.16 USD
77339-956 77347-480 77371-482
5-Bromo-4-methoxy-2-methylphenol ≥95%
5-Bromo-4-methoxy-2-methylphenol

Precaución: for Research Use Only

Formula: C₈H₉BrO₂
Storage Temperature: Ambient
MDL Number: MFCD24722518
CAS Number: 14078-73-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR