3-Chloro-2-(1-piperidinyl)-5-(trifluoromethyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Chloro-2-(piperidin-1-yl)-5-(trifluoromethyl)pyridine

107392-500MG 107392-1G 107392-5G 107392-25G
77323-416EA 95.79 USD
77323-416 77342-600 77359-512 77371-474
3-Chloro-2-(1-piperidinyl)-5-(trifluoromethyl)pyridine ≥97%
3-Chloro-2-(1-piperidinyl)-5-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₁₁H₁₂ClF₃N₂
MW: 264.68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13561834
CAS Number: 1219967-90-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR