3-Chloro-2-(1-pyrrolidinyl)-5-(trifluoromethyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Chloro-2-(pyrrolidin-1-yl)-5-(trifluoromethyl)pyridine

107391-500MG 107391-1G 107391-5G
77338-862EA 526.33 USD
77338-862 77347-476 77371-472
3-Chloro-2-(1-pyrrolidinyl)-5-(trifluoromethyl)pyridine ≥97%
3-Chloro-2-(1-pyrrolidinyl)-5-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₁₀H₁₀ClF₃N₂
MW: 250.65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13561832
CAS Number: 122599-20-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR