1-Chloro-2-iodo-3-methyl-5-nitrobenzene ≥95%

Supplier: AOBChem USA

Warning

107327-500MG 107327-1G 107327-5G 107327-25G
77323-412EA 190.55 USD
77323-412 77347-474 77359-508 77383-442
1-Chloro-2-iodo-3-methyl-5-nitrobenzene ≥95%
1-Chloro-2-iodo-3-methyl-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₇H₅ClINO₂
Storage Temperature: Ambient
MDL Number: MFCD01313705
CAS Number: 959575-00-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR