1,2-Dichloro-3-methyl-5-nitrobenzene ≥95%

Supplier: AOBChem USA

Warning

107325-500MG 107325-1G 107325-5G
77323-410EA 334.75 USD
77323-410 77347-472 77371-470
1,2-Dichloro-3-methyl-5-nitrobenzene ≥95%
1,2-Dichloro-3-methyl-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₇H₅Cl₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD12974172
CAS Number: 51719-46-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR