3-Bromo-4-fluoro-5-methylbenzonitrile ≥97%

Supplier: AOBChem USA

Warning

107094-500MG 107094-1G 107094-5G 107094-25G
77323-408EA 276.04 USD
77323-408 77347-470 77359-502 77383-440
3-Bromo-4-fluoro-5-methylbenzonitrile ≥97%
3-Bromo-4-fluoro-5-methylbenzonitrile

Precaución: for Research Use Only

Formula: C₈H₅BrFN
MW: 214.034 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28739739
CAS Number: 1805108-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR