(4-Fluoro-2,6-dimethylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

106896-500MG 106896-1G 106896-25G 106896-5G
77323-406EA 287.37 USD
77323-406 77347-468 77383-438 77371-464
(4-Fluoro-2,6-dimethylphenyl)(methyl)sulfane ≥97%
(4-Fluoro-2,6-dimethylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₁FS
MW: 170.247 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00464331
CAS Number: 38380-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR