5-Borono-2-fluoro-4-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

106778-500MG 106778-1G 106778-5G 106778-25G
77323-404EA 277.07 USD
77323-404 77347-466 77359-498 77383-436
5-Borono-2-fluoro-4-methylbenzoic acid ≥97%
5-Borono-2-fluoro-4-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₈BFO₄
MW: 197.956 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34779198
CAS Number: 1952356-90-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR