4-Amino-3-bromo-5-fluorobenzotrifluoride ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-6-fluoro-4-(trifluoromethyl)aniline

106565-500MG 106565-1G 106565-5G 106565-25G
77323-400EA 282.22 USD
77323-400 77347-462 77359-496 77383-434
4-Amino-3-bromo-5-fluorobenzotrifluoride ≥97%
4-Amino-3-bromo-5-fluorobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₄BrF₄N
MW: 258.011 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28397844
CAS Number: 1034325-63-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR