2,4-Dibromo-6-chloro-3-(trifluoromethyl)aniline ≥97%

Supplier: AOBChem USA

Warning

106317-500MG 106317-1G 106317-5G
77338-584EA 501.61 USD
77338-584 77347-458 77371-458
2,4-Dibromo-6-chloro-3-(trifluoromethyl)aniline ≥97%
2,4-Dibromo-6-chloro-3-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₇H₃Br₂ClF₃N
MW: 353.362 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30712687
CAS Number: 1335044-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR