N-Methoxy-N,4-dimethylthiophene-2-carboxamide ≥97%

Supplier: AOBChem USA

Warning

106287-500MG 106287-1G 106287-5G 106287-25G
77323-396EA 202.91 USD
77323-396 77347-456 77359-492 77383-430
N-Methoxy-N,4-dimethylthiophene-2-carboxamide ≥97%
N-Methoxy-N,4-dimethylthiophene-2-carboxamide

Precaución: for Research Use Only

Formula: C₈H₁₁NO₂S
MW: 185.243 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28165897
CAS Number: 1862972-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR