4-Methylthiophene-2-carboxamide ≥97%

Supplier: AOBChem USA

Warning

106282-1G 106282-5G 106282-25G
77341-648EA 97.85 USD
77341-648 77359-490 77371-456
4-Methylthiophene-2-carboxamide ≥97%
4-Methylthiophene-2-carboxamide

Precaución: for Research Use Only

Formula: C₆H₇NOS
Storage Temperature: Ambient
MDL Number: MFCD06657971
CAS Number: 83933-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR