1-(4-Bromo-2-isopropylphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

105820-500MG 105820-1G 105820-5G 105820-25G
77323-392EA 120.51 USD
77323-392 77343-654 77359-486 77371-454
1-(4-Bromo-2-isopropylphenyl)ethanone ≥97%
1-(4-Bromo-2-isopropylphenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrO
MW: 241.124 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32266622
CAS Number: 2403679-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR