4-Acetyl-2-methoxybenzoic acid ≥95%

Supplier: AOBChem USA

Warning

105729-500MG 105729-1G 105729-5G 105729-25G
77323-390EA 133.9 USD
77323-390 77344-492 77359-484 77383-426
4-Acetyl-2-methoxybenzoic acid ≥95%
4-Acetyl-2-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₄
Storage Temperature: Ambient
MDL Number: MFCD20644542
CAS Number: 102362-04-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR