(4-Acetyl-2-methoxyphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

105727-500MG 105727-1G 105727-5G 105727-25G
77323-388EA 280.16 USD
77323-388 77347-452 77359-482 77383-424
(4-Acetyl-2-methoxyphenyl)boronic acid ≥97%
(4-Acetyl-2-methoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BO₄
Storage Temperature: Ambient
MDL Number: MFCD34778589
CAS Number: 1992804-71-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR